2-Aminoazotoluol 97%
Lieferant: Thermo Scientific
|
Gefahr
|
153260250
ACRO153260250EA
42
EUR
ACRO153260250
2-Aminoazotoluol 97%
2-Aminoazotoluol
Formel:
C₁₄H₁₅N₃ Molecular Weight: 225,29 g/mol Schmelzpunkt: 101…102 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007733 CAS-Nummer: 97-56-3 EINECS: 202-591-2 |
Spezifikation Testergebnisse
Appearance | Orange to red-brown to dark brown Powder |
Infrared spectrum | Conforms |
Melting point | 100 to 104 °C |
Loss on drying | ≤3.0 % (110 °C, 1 h) |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...