3,5-Dibromsalicylaldehyd 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
153400250
153401000
ACRO153400250EA
122
EUR
ACRO153400250
ACRO153401000
3,5-Dibromsalicylaldehyd 98%
3,5-Dibromsalicylaldehyd
Formel:
C₇H₄Br₂O₂ Siedepunkt: 261 °C (1013 hPa) Schmelzpunkt: 80…84 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003318 CAS-Nummer: 90-59-5 EINECS: 202-003-4 Merck Index: 13,03052 |
Spezifikation Testergebnisse
Appearance | Yellow to yellow-brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 80 to 84 °C |
Titration with NaOH | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...