Benzo[h]chinolin 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 7,8-Benzoquinoline, alpha-Naphthoquinoline, 4-Azaphenanthrene

154070010
ACRO154070010EA 31.2 EUR
ACRO154070010
Benzo[h]chinolin 97%
Benzo[h]chinolin

Appearance: White to brown Crystalline powder or crystals and/or chunks

Formel: C₁₃H₉N
Molecular Weight: 179.22 g/mol
Siedepunkt: 338 °C (719 mmHg)
Schmelzpunkt: 48…52 °C
Flammpunkt: >112 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004984
CAS-Nummer: 230-27-3

Bestellen

Spezifikation Testergebnisse

Appearance White to brown Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 48 to 53 °C
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR