α,α'-Dichlor-o-xylol 98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
1,2-Bis(chlormethyl)benzol, o-Xyloldichlorid, o-Xylylendichlorid
154141000
ACRO154141000EA
125
EUR
ACRO154141000
α,α'-Dichlor-o-xylol 98%
α,α'-Dichlor-o-xylol
Formel:
C₈H₈Cl₂ Molecular Weight: 175,06 g/mol Siedepunkt: 240 °C (1013 hPa) Schmelzpunkt: 53…57 °C Dichte: 1,393 g/cm³ (0 °C) Flammpunkt: 107 °C |
MDL:
MFCD00000903 CAS-Nummer: 612-12-4 EINECS: 210-291-8 UN: 2928 ADR: 6.1,II |
Spezifikation Testergebnisse
Appearance | Semitransparent white to light beige Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 53 to 57 °C |
GC | ≥97.5 % |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...