5-Bromisatin 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 5-Brom-2,3-indolindion, 5-Brom-1H-indol-2,3-dion

154230050 154230250
ACRO154230050EA 27.3 EUR
ACRO154230050 ACRO154230250
5-Bromisatin 98%
5-Bromisatin
Formel: C₈H₄BrNO₂
Molecular Weight: 226.03 g/mol
Schmelzpunkt: 251…253 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00149345
CAS-Nummer: 87-48-9

Bestellen

Spezifikation Testergebnisse

Appearance Orange Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
GC ≥96 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR