5-Bromisatin 98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
5-Brom-2,3-indolindion, 5-Brom-1H-indol-2,3-dion
154230050
154230250
ACRO154230050EA
27.3
EUR
ACRO154230050
ACRO154230250
5-Bromisatin 98%
5-Bromisatin
Formel:
C₈H₄BrNO₂ Molecular Weight: 226.03 g/mol Schmelzpunkt: 251…253 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149345 CAS-Nummer: 87-48-9 |
Spezifikation Testergebnisse
Appearance | Orange Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
GC | ≥96 % |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...