Bis-(cyclohexanon)-oxaldihydrazon 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Cupferazone, N'1,N'2-Dicyclohexylideneoxalohydrazide, Oxalic acid bis(cyclohexylidenehydrazide), Bis(cyclohexanone)oxaldihydrazone, Cuprizone

154420250 154421000
ACRO154420250EA 62.6 EUR
ACRO154420250 ACRO154421000
Bis-(cyclohexanon)-oxaldihydrazon 98%
Bis(cyclohexanone)oxalyldihydrazone

Appearance: White Powder

Formel: C₁₄H₂₂N₄O₂
Schmelzpunkt: 208…214 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00001659
CAS-Nummer: 370-81-0
UN: 2811
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance White to almost white Powder
Infrared spectrum Conforms
Melting point 208 to 214 °C
Loss on drying ≤0.5 %
Sulfated ash ≤0.1 %
Titration with NaOH 97.5 to 102.5 %
Suitability (for Cu determination) Passes test
Iron (Fe) ≤10 ppm
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR