N-(2-Cyanethyl)glycin 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
N-(2-Cyanoethyl)aminoacetic Acid
154901000
ACRO154901000EA
222
EUR
ACRO154901000
N-(2-Cyanethyl)glycin 98%
N-(2-Cyanethyl)glycin
Appearance: White Powder or flakes
Formel:
C₅H₈N₂O₂ Molecular Weight: 128,13 g/mol Schmelzpunkt: 188…193 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00066903 CAS-Nummer: 3088-42-4 UN: 3439 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | White Powder or flakes |
Infrared spectrum | Conforms |
Melting point | 188 to 193 °C |
Titration with HClO4 | ≥97.5 % |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...