trans-1,2-Diaminocyclohexan-N,N,N',N'-tetraessigsäure Monohydrat 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
155000250
155001000
155000025
155005000
ACRO155000250EA
72
EUR
ACRO155000250
ACRO155001000
ACRO155000025
ACRO155005000
trans-1,2-Diaminocyclohexan-N,N,N',N'-tetraessigsäure Monohydrat 98%
trans-1,2-Diaminocyclohexan-N,N,N',N'-tetraessigsäure Monohydrat
Formel:
C₁₄H₂₂N₂O₈·1H₂O Schmelzpunkt: >210 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00066429 CAS-Nummer: 125572-95-4 EINECS: 236-308-9 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Titration Complexometric | 97.5 to 102.5 % |
Loss on drying | 4.0 to 6.9 % (1g,150°C,6h) |
Sulfated ash | ≤0.3 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...