N,N'-Diethylethylendiamin 95%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
155520050
155520250
ACRO155520050EA
33.9
EUR
ACRO155520050
ACRO155520250
N,N'-Diethylethylendiamin 95%
N,N'-Diethylethylendiamin
Formel:
C₆H₁₆N₂ Siedepunkt: 152…154 °C (760 mmHg) Schmelzpunkt: –11 °C Dichte: 0,811 g/cm³ Flammpunkt: 33 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009033 CAS-Nummer: 111-74-0 UN: 2685 ADR: 8,II |
Spezifikation Testergebnisse
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4315 to 1.4335 (20°C, 589 nm) |
GC | ≥94 % |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...