N-Ethylethylendiamin 98+%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
2-Aminoethyl(ethyl)amin, 1-Amino-2-(ethylamino)ethan
155550500
ACRO155550500EA
139
EUR
ACRO155550500
N-Ethylethylendiamin 98+%
N-Ethylethylendiamin
Formel:
C₄H₁₂N₂ Molecular Weight: 88.15 g/mol Siedepunkt: 128 Dichte: 0.83 g/cm³ Flammpunkt: 10 °C |
MDL:
MFCD00008166 CAS-Nummer: 110-72-5 EINECS: 203-795-4 UN: 2733 ADR: 3 II |
Spezifikation Testergebnisse
Appearance | Clear colorless to light brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4380 to 1.4400 (20°C, 589 nm) |
GC | ≥98.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...