2-Amino-5-bromothiazole hydrobromide 95%
Lieferant: Thermo Scientific
Synonyme:
2-Amino-5-bromothiazole monohydrobromide, 2-Amino-5-bromothiazole HBr
156000250
ACRO156000250EA
149
EUR
ACRO156000250
2-Amino-5-bromothiazole hydrobromide 95%
2-Amino-5-bromothiazole hydrobromide
Appearance: Light yellow to light brown Crystalline powder
Formel:
C₃H₃BrN₂S Molecular Weight: 259,95 g/mol Schmelzpunkt: 165 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012712 CAS-Nummer: 61296-22-8 |
Spezifikation Testergebnisse
Appearance | Light yellow to light brown Crystalline powder |
Infrared spectrum | Conforms |
TLC | ≥94 % |
Titration with HClO4 | ≥94 % |
GC | plus methylcyclopentane) |
UV | at 420 nm A: ≤0.005 |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.0005 % |
Acidity | ≤0.0002 meq/g |
Sulfur compounds (as S) | ≤0.005 % |
Water soluble titr. acid | ≤0.0003 meq/g |
Thiophene | Passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...