Diethyl furan-3,4-dicarboxylate 95%
Lieferant: Thermo Scientific
Achtung
|
Synonyme:
Diethyl 3,4-furandicarboxylate
156510050
ACRO156510050EA
115
EUR
ACRO156510050
Diethyl furan-3,4-dicarboxylate 95%
Diethyl furan-3,4-dicarboxylate
Formel:
C₁₀H₁₂O₅ Molecular Weight: 212,2 g/mol Siedepunkt: 155 °C (13 mmHg) Dichte: 1,14 g/cm³ Flammpunkt: 82 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005349 CAS-Nummer: 30614-77-8 |
Spezifikation Testergebnisse
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4710 to 1.4730 (20°C, 589 nm) |
GC | ≥99.4 % |
Color scale | ≤10 APHA |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...