4-Methylaminophenolsulfat 99%, reinst

Lieferant: Thermo Fisher Scientific
Achtung

156800050 156801000 156805000 156800025
ACRO156800050EA 26.4 EUR
ACRO156800050 ACRO156801000 ACRO156805000 ACRO156800025
4-Methylaminophenolsulfat 99%, reinst
4-Methylaminophenolsulfat
Formel: C₁₄H₂₀N₂O₆S
Siedepunkt: 227 °C (1013 hPa)
Schmelzpunkt: 260 °C (Zers.)
Dichte: 1,434 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00013140
CAS-Nummer: 55-55-0
EINECS: 200-237-1
UN: 2811
ADR: 6.1,III
Merck Index: 13,06046

Bestellen

Spezifikation Testergebnisse

Appearance White to beige to grey Crystalline powder or crystals
Infrared spectrum Conforms
Loss on drying ≤0.3 %
Residue after ignition ≤0.15 %
Titration with NaOH ≥98.5 %
Iron (Fe) ≤50 ppm
Heavy metals (as Pb) ≤30 ppm
Potassium (K) ≤0.002 %
Sodium (Na) ≤0.005 %
Heavy metals (ICP-OES) ≤0.001 %
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR