N-Hydroxysuccinimid 98+%

Lieferant: Thermo Fisher Scientific
Achtung

157270250 157271000 157272500 157270010
ACRO157270250EA 38.4 EUR
ACRO157270250 ACRO157271000 ACRO157272500 ACRO157270010
N-Hydroxysuccinimid 98+%
N-Hydroxysuccinimid
Formel: C₄H₅NO₃
Siedepunkt: 363 °C (1013 hPa)
Schmelzpunkt: 93…95 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005516
CAS-Nummer: 6066-82-6
EINECS: 228-001-3

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder or crystals (May have a tendency to cake)
Infrared spectrum Conforms
Water ≤2.0 % (K.F.)
Solubility The product may contain some hydroxysuccinimide:ammonia adduct, which is virtually insoluble in acetone
Titration with NaOH 98.0 to 101.0 % (On dried substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR