Indigocarmin, hochrein biologische Färbemittel, zur Analyse
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Acid Blue 74, Food blue No. 2
157440250
157441000
157445000
ACRO157440250EA
39.5
EUR
ACRO157440250
ACRO157441000
ACRO157445000
Indigocarmin, hochrein biologische Färbemittel, zur Analyse
Indigocarmin
Formel:
C₁₆H₈N₂Na₂O₈S₂ Schmelzpunkt: ? 300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005723 CAS-Nummer: 860-22-0 EINECS: 212-728-8 Merck Index: 13,04969 |
Spezifikation Testergebnisse
Appearance | Dark blue Crystalline powder |
Infrared spectrum | Conforms |
Heavy metals | ≤40 ppm |
Additional info | dinatrium-3,3'-dioxo-2,2'-bisindolylidene-5,7'-disulphonate: 18.0% |
Dye content | ≥85 % |
Arsenic (As) | ≤3 ppm |
Cadmium (Cd) | ≤1 ppm |
Lead (Pb) | ≤10 ppm |
Mercury (Hg) | ≤1 ppm |
Free amine | ≤0.01 % (aromatic) |
Insoluble matter | ≤0.2 % (in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...