Indigocarmin, hochrein biologische Färbemittel, zur Analyse

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Acid Blue 74, Food blue No. 2

157440250 157441000 157445000
ACRO157440250EA 39.5 EUR
ACRO157440250 ACRO157441000 ACRO157445000
Indigocarmin, hochrein biologische Färbemittel, zur Analyse
Indigocarmin
Formel: C₁₆H₈N₂Na₂O₈S₂
Schmelzpunkt: ? 300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005723
CAS-Nummer: 860-22-0
EINECS: 212-728-8
Merck Index: 13,04969

Bestellen

Spezifikation Testergebnisse

Appearance Dark blue Crystalline powder
Infrared spectrum Conforms
Heavy metals ≤40 ppm
Additional info dinatrium-3,3'-dioxo-2,2'-bisindolylidene-5,7'-disulphonate: 18.0%
Dye content ≥85 %
Arsenic (As) ≤3 ppm
Cadmium (Cd) ≤1 ppm
Lead (Pb) ≤10 ppm
Mercury (Hg) ≤1 ppm
Free amine ≤0.01 % (aromatic)
Insoluble matter ≤0.2 % (in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR