1,2-Phenylendiessigsäure 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
157550050
157550250
ACRO157550050EA
41.6
EUR
ACRO157550050
ACRO157550250
1,2-Phenylendiessigsäure 99%
1,2-Phenylendiessigsäure
Formel:
C₁₀H₁₀O₄ Schmelzpunkt: 150…152 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004329 CAS-Nummer: 7500-53-0 EINECS: 231-355-1 |
Spezifikation Testergebnisse
Appearance | Beige to light brown Crystalline powder or needles |
Infrared spectrum | Conforms |
Melting point | 150 to 152 °C |
Titration with NaOH | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...