Pyrazinamid 99%

Lieferant: Thermo Fisher Scientific

157640250 157641000 157640100
ACRO157640250EA 75.4 EUR
ACRO157640250 ACRO157641000 ACRO157640100
Pyrazinamid 99%
Pyrazinamid
Formel: C₅H₅N₃O
Siedepunkt: 357 °C (1013 hPa)
Schmelzpunkt: 188…191 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006132
CAS-Nummer: 98-96-4
EINECS: 202-717-6
Merck Index: 12,08140

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 188 to 191 °C
HPLC ≥98.5 %
Iodate (IO3) ≤0.0005 %
Nitrite (NO2) ≤0.001 %
Phosphate (PO4) ≤0.0005 %
Sulfate (SO4) ≤0.003 %
Calcium (Ca) ≤0.005 %
Iron (Fe) ≤0.0003 %
Magnesium (Mg) ≤0.002 %
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR