1,2-Diaminocyclohexan (Gemisch der cis- und trans-Isomeren) cis- and trans- mixture 99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
1,2-Diaminocyclohexane, 1,2-Cyclohexanediamine
158100010
158102500
ACRO158100010EA
113
EUR
ACRO158100010
ACRO158102500
1,2-Diaminocyclohexan (Gemisch der cis- und trans-Isomeren) cis- and trans- mixture 99%
1,2-Diaminocyclohexan (Gemisch der cis- und trans-Isomeren)
Formel:
C₆H₁₄N₂ Molecular Weight: 114.19 g/mol Siedepunkt: 192 Schmelzpunkt: 2…15 °C Dichte: 0.93 g/cm³ Flammpunkt: 75 °C |
MDL:
MFCD00001491 CAS-Nummer: 694-83-7 EINECS: 211-776-7 UN: 2735 |
Spezifikation Testergebnisse
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4860 to 1.4920 (20°C, 589 nm) |
GC | ≥98.5 % |
Color scale | ≤75 APHA |
Titration with HCl | 99.4 to 100.6 % |
Chloride (Cl) | ≤20 ppm |
Sulfate (SO4) | ≤150 ppm |
Ammonium (NH4) | ≤5 ppm |
Calcium (Ca) | ≤100 ppm |
Copper (Cu) | ≤2 ppm |
Iron (Fe) | ≤5 ppm |
Magnesium (Mg) | ≤50 ppm |
Potassium (K) | ≤50 ppm |
Zinc (Zn) | ≤5 ppm |
Heavy metals (as Pb) | ≤5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...