trans-2-Hexenal 99%

Lieferant: Thermo Fisher Scientific
Gefahr

158130250 158131000
ACRO158130250EA 38.4 EUR
ACRO158130250 ACRO158131000
trans-2-Hexenal 99%
trans-2-Hexenal
Formel: C₆H₁₀O
Siedepunkt: 147 °C (1013 hPa)
Dichte: 0,846 g/cm³
Flammpunkt: 35 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00007008
CAS-Nummer: 6728-26-3
EINECS: 229-778-1
UN: 1988
ADR: 3,III

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4440 to 1.4500 (20°C, 589 nm)
Density (20°C) 0.843 to 0.849 g/ml
GC ≥98.5 %
Acid value ≤5 mg KOH/g
Chloride (Cl) ≤5 ppm
Nitrate (NO3) ≤10 ppm
Phosphate (PO4) ≤5 ppm
Arsenic (As) ≤1 ppm
Cadmium (Cd) ≤1 ppm
Calcium (Ca) ≤10 ppm
Copper (Cu) ≤1 ppm
Iron (Fe) ≤2 ppm
Zinc (Zn) ≤1 ppm
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR