Pyridiniumtribromid 90+%, technische Qualität

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Pyridinium bromide perbromide, Monopyridin-1-ium tribromide, Pyridine hydrobromide perbromide

158361000 158365000
ACRO158361000EA 118 EUR
ACRO158361000 ACRO158365000
Pyridiniumtribromid 90+%, technische Qualität
Pyridinium tribromide
Formel: C₅H₆Br₃N
Molecular Weight: 319.82 g/mol
Schmelzpunkt: 127…133 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00013223
CAS-Nummer: 39416-48-3
EINECS: 254-446-8
UN: 3261
ADR: 8 II

Bestellen

Spezifikation Testergebnisse

Appearance Orange to red to brown Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 130 to 137 °C (Dec)
Titration Iodometric ≥90.0 %
UV (0.1 M in water)
pH 3.8 to 4.5 (1 M in water)
Titration with NaOH ≥99.4 %
Chloride (Cl) ≤0.005 %
Iron (Fe) ≤0.0005 %
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR