2-Amino-5-methyl-1,3,4-thiadiazol 97%

Lieferant: Thermo Scientific

158370050
ACRO158370050EA 43.9 EUR
ACRO158370050
2-Amino-5-methyl-1,3,4-thiadiazol 97%
2-Amino-5-methyl-1,3,4-thiadiazole
Formel: C₃H₅N₃S
Molecular Weight: 115,16 g/mol
Schmelzpunkt: 224…225 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003110
CAS-Nummer: 108-33-8
EINECS: 203-573-7

Bestellen

Spezifikation Testergebnisse

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥96.0 %
Acidity Passes test
Alkalinity Passes test
Sulfate (SO4) ≤0.03 %
Aluminium (Al) ≤1 ppm
Iron (Fe) ≤10 ppm
Heavy metals (as Pb) ≤10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR