N-Boc-glycin (Boc-Gly-OH) 99+%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Boc-Gly-OH

158800100 158801000
ACRO158800100EA 42 EUR
ACRO158800100 ACRO158801000
N-Boc-glycin (Boc-Gly-OH) 99+%
N-Boc-glycin (Boc-Gly-OH)
Formel: C₇H₁₃NO₄
Schmelzpunkt: 87…90 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00002690
CAS-Nummer: 4530-20-5
EINECS: 224-864-5

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 85 to 90 °C
Water ≤0.5 % (K.F.)
HPLC ≥99.0 % (ELSD)
Titration with NaOH ≥99.0 %
Heavy metals (as Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR