3-(4,5-Dimethyl-2-thiazolyl)-2,5-diphenyltetrazoliumbromid 98%

Lieferant: Thermo Fisher Scientific
Achtung

158990010 158990050 158992500
ACRO158990010EA 51.2 EUR
ACRO158990010 ACRO158990050 ACRO158992500
3-(4,5-Dimethyl-2-thiazolyl)-2,5-diphenyltetrazoliumbromid 98%
3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT)
Formel: C₁₈H₁₆N₅BrS
Lagertemperatur: Kühlschrank
MDL: MFCD00011964
CAS-Nummer: 298-93-1
EINECS: 206-069-5

Bestellen

Spezifikation Testergebnisse

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %
Specific extinction E (1%/1cm) ≥480 (243 nm, water)
Chloride (Cl) ≤0.002 %
Nitrite (NO2) ≤0.001 %
Phosphate (PO4) ≤0.003 %
Sulfate (SO4) ≤0.015 %
Calcium (Ca) ≤0.01 %
Iron (Fe) ≤0.002 %
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR