Isochinolin-5-amin 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Isoquinolin-5-amine, 5-Aminoisoquinoline
159420010
159420050
ACRO159420010EA
32
EUR
ACRO159420010
ACRO159420050
Isochinolin-5-amin 99%
Isochinolin-5-amin
Formel:
C₉H₈N₂ Schmelzpunkt: 127…130 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006907 CAS-Nummer: 1125-60-6 EINECS: 214-408-3 UN: 2811 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | Beige to yellow-brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 126 to 130 °C |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...