2-Methyl-4(5)-nitroimidazol 99%

Lieferant: Thermo Fisher Scientific

Synonyme: 2-Methyl-4-nitroimidazol, 2-Methyl-4-nitro-1H-imidazol

159470500 159472500
ACRO159470500EA 36.9 EUR
ACRO159470500 ACRO159472500
2-Methyl-4(5)-nitroimidazol 99%
2-Methyl-4(5)-nitroimidazol
Formel: C₄H₅N₃O
Molecular Weight: 127.10 g/mol
Schmelzpunkt: 252…254 °C
MDL: MFCD00005191
CAS-Nummer: 88054-22-2
EINECS: 211-790-3

Bestellen

Spezifikation Testergebnisse

Appearance White to beige Powder
Infrared spectrum Conforms
Titration with HClO4 98.0 to 102.0 % (On dried substance)
Sodium dihydrogenphosphate ≤2.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR