Dimethylmalonat 97%

Lieferant: Thermo Fisher Scientific

Synonyme: 1,3-Propanedioic acid dimethyl ester, Malonic acid dimethyl ester

159560010 159562500 159560025
ACRO159560010EA 88.9 EUR
ACRO159560010 ACRO159562500 ACRO159560025
Dimethylmalonat 97%
Dimethylmalonat
Formel: C₅H₈O₄
Molecular Weight: 132.12 g/mol
Siedepunkt: 180…181 °C (760 mmHg)
Schmelzpunkt: –62 °C
Dichte: 1.156 g/cm³
Flammpunkt: 90 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008460
CAS-Nummer: 108-59-8
EINECS: 203-597-8
Merck Index: 12,06169

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4120 to 1.4140 (20°C, 589 nm)
GC ≥96.0 %
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR