Dimethylmalonat 97%
Lieferant: Thermo Fisher Scientific
Synonyme:
1,3-Propanedioic acid dimethyl ester, Malonic acid dimethyl ester
159560010
159562500
159560025
ACRO159560010EA
88.9
EUR
ACRO159560010
ACRO159562500
ACRO159560025
Dimethylmalonat 97%
Dimethylmalonat
Formel:
C₅H₈O₄ Molecular Weight: 132.12 g/mol Siedepunkt: 180…181 °C (760 mmHg) Schmelzpunkt: –62 °C Dichte: 1.156 g/cm³ Flammpunkt: 90 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008460 CAS-Nummer: 108-59-8 EINECS: 203-597-8 Merck Index: 12,06169 |
Spezifikation Testergebnisse
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4120 to 1.4140 (20°C, 589 nm) |
GC | ≥96.0 % |
Heavy metals (as Pb) | ≤10 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...