1-(3-aminopropyl)pyrrolidin-2-one 95%

Lieferant: Thermo Scientific
Gefahr

Synonyme: N-(3'-Aminopropyl)-2-pyrrolidinone, 1-(3-Aminopropyl)pyrrolidin-2-one

159600050
ACRO159600050EA 40.2 EUR
ACRO159600050
1-(3-aminopropyl)pyrrolidin-2-one 95%
1-(3-aminopropyl)pyrrolidin-2-one

Appearance: Clear colorless to light brown Liquid

Formel: C₇H₁₄N₂O
Molecular Weight: 142,202 g/mol
Siedepunkt: 120…123 °C (1 mmHg)
Dichte: 1,01 g/cm³
Flammpunkt: >112 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003201
CAS-Nummer: 7663-77-6
UN: 3267
ADR: 8,II

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.4970 to 1.4990 (20°C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR