n-Hexan 99+% zur Analyse
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
160780010
160780025
160780251
ACRO160780010EA
113
EUR
ACRO160780010
ACRO160780025
ACRO160780251
n-Hexan 99+% zur Analyse
n-Hexan
Formel:
H₃C(CH₂)₄CH₃ Siedepunkt: 69 °C (760 mmHg) Schmelzpunkt: –95 °C Dichte: 0,659 g/cm³ Flammpunkt: –22 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009520 CAS-Nummer: 110-54-3 EINECS: 203-777-6 UN: 1208 ADR: 3,II Merck Index: 13,04712 |
|
Spezifikation Testergebnisse
Appearance | Clear liquid |
Water | ≤0.02 % (Coulometric) |
GC | ≥99.0 % |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.002 % |
Calcium (Ca) | ≤0.5 ppm |
Copper (Cu) | ≤0.1 ppm |
Iron (Fe) | ≤0.1 ppm |
Lead (Pb) | ≤0.1 ppm |
Magnesium (Mg) | ≤0.1 ppm |
Zinc (Zn) | ≤0.1 ppm |
Sulfur compounds (as S) | ≤0.005 % |
Free acid (CH3COOH) | ≤0.002 % |
Thiophene | Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...