N,N-Dimethylformamiddimethylacetal ca. 97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 1,1-Dimethoxytrimethylamin

161251000 161255000 161250250
ACRO161251000EA 200 EUR
ACRO161251000 ACRO161255000 ACRO161250250
N,N-Dimethylformamiddimethylacetal ca. 97%
N,N-Dimethylformamiddimethylacetal
Formel: C₅H₁₃NO₂
Siedepunkt: 102 °C (1013 hPa)
Dichte: 0,89 g/cm³
Flammpunkt: 7 °C
MDL: MFCD00008482
CAS-Nummer: 4637-24-5
EINECS: 225-063-3
UN: 1993

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3950 to 1.3970 (20°C, 589 nm)
GC ≥96.0 %
Specific optical rotation -53° to -63° (20°C, 589 nm) (c=0.5, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR