4-Amino-m-kresol 99+%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
163200500
163202500
ACRO163200500EA
37.6
EUR
ACRO163200500
ACRO163202500
4-Amino-m-kresol 99+%
4-Amino-m-kresol
Formel:
C₇H₉NO Siedepunkt: 259 °C (1013 hPa) Schmelzpunkt: 176…180 °C |
MDL:
MFCD00007871 CAS-Nummer: 2835-99-6 EINECS: 220-621-2 |
Spezifikation Testergebnisse
Appearance | Off-white to yellow to beige to brown Powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 176 to 180 °C |
GC | ≥99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...