3-Methoxy-2(1H)-pyridone 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
163540010
163540050
ACRO163540010EA
75.6
EUR
ACRO163540010
ACRO163540050
3-Methoxy-2(1H)-pyridone 97%
3-Methoxy-2(1H)-pyridone
Formel:
C₆H₇NO₂ Schmelzpunkt: 113…115 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006015 CAS-Nummer: 20928-63-6 |
Spezifikation Testergebnisse
Appearance | White to brown Powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...