Chininsulfat Dihydrat 99+%

Lieferant: Thermo Fisher Scientific
Achtung

163710100 163710500
ACRO163710100EA 59.5 EUR
ACRO163710100 ACRO163710500
Chininsulfat Dihydrat 99+%
Chininsulfat Dihydrat
Formel: C₄₀H₅₀N₄O₈S·2H₂O
Schmelzpunkt: 233…235 °C
Dichte: 1,36 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00150790
CAS-Nummer: 6119-70-6
EINECS: 212-359-2
Merck Index: 12,03822

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder or crystals or needles
Infrared spectrum Conforms
Water 4.0 to 5.5 % (K.F.)
Residue after ignition ≤0.1 %
Specific optical rotation -235° to -245° (20°C, 589 nm) (c=2, 0.1N HCl) (on anhydrous basis)
Titration with HClO4 99.0 to 101.0 % (On anhydrous basis)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR