(-)-Chinin Hydrochlorid Dihydrat 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Quinine hydrochloride dihydrate, Quinine monohydrochloride dihydrate, (R)-(6-Methoxyquinolin-4-yl)((1S,2S,4S,5R)-5-vinylquinuclidin-2-yl)methanol hydrochloride dihydrate

163720250 163720050 163721000
ACRO163720250EA 75.5 EUR
ACRO163720250 ACRO163720050 ACRO163721000
(-)-Chinin Hydrochlorid Dihydrat 99%
(-)-Chinin Hydrochlorid Dihydrat
Formel: C₂₀H₂₅ClN₂O₂·2H₂O
Molecular Weight: 396.91 g/mol
Schmelzpunkt: 115…116 °C (Zers.)
Flammpunkt: 122 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00151248
CAS-Nummer: 6119-47-7
EINECS: 205-001-1

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder or needles
Infrared spectrum Conforms
Loss on drying 6 % to 10 % (1 g, 105°C)
Sulfated ash ≤0.1 %
Specific optical rotation -245° to -258° (20°C, 589 nm) (c=2, 0.1 M HCl) (on dried substance)
Titration with HClO4 ≥98.5 %
Sulfate (SO4) ≤500 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR