Adenosin 99+%
Lieferant: Thermo Fisher Scientific
Synonyme:
9-β-D-Ribofuranosyladenine, (2R,3R,4S,5R)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, Adenine-9-β-D-ribofuranoside, Adenine riboside
164040050
164040250
164041000
164045000
ACRO164040050EA
32.7
EUR
ACRO164040050
ACRO164040250
ACRO164041000
ACRO164045000
Adenosin 99+%
Adenosin
Formel:
C₁₀H₁₃N₅O₄ Molecular Weight: 267.24 g/mol Schmelzpunkt: 233…238 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005752 CAS-Nummer: 58-61-7 EINECS: 200-389-9 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 233 to 238 °C |
Loss on drying | ≤0.5 % (105 °C, 2 hrs) |
Sulfated ash | ≤0.1 % |
HPLC | ≥99.0 % |
UV spectrophotometry | molar absorptivity 15400 +/- 2% , on dry basis |
UV transmittance | ≥98 % (c=0.5, H2O, 430 nm) 1 cm cell |
Ratio of Absorbances | 0.14 to 0.16 (A280/A260) |
Nitrogen (N) | 25.69 to 26.73 % (on dry basis) |
Heavy metals (as Pb) | ≤10 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...