Bathocuproinsulfonat Dinatriumsalz Hydrat 97%

Lieferant: Thermo Fisher Scientific

164060010 164060050 164060250
ACRO164060010EA 108 EUR
ACRO164060010 ACRO164060050 ACRO164060250
Bathocuproinsulfonat Dinatriumsalz Hydrat 97%
Bathocuproinsulfonat Dinatriumsalz Hydrat
Formel: C₂₆H₁₈N₂Na₂O₆S₂·H₂O
Schmelzpunkt: 300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00149974
CAS-Nummer: 52698-84-7
EINECS: 258-111-7

Bestellen

Spezifikation Testergebnisse

Appearance White to beige Powder
Infrared spectrum Conforms
Loss on drying ≤9 %
HPLC ≥96.0 % (expressed as the sum of ortho-ortho, meta-para and para-para-substituted bathocuproine disulfonate)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR