4-Amino-2-nitrophenol 99%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 4-Hydroxy-3-nitroanilin

164230050
ACRO164230050EA 33.3 EUR
ACRO164230050
4-Amino-2-nitrophenol 99%
4-Amino-2-nitrophenol
Formel: C₆H₆N₂O₃
Molecular Weight: 154.13 g/mol
Schmelzpunkt: 124…128 °C
MDL: MFCD00007876
CAS-Nummer: 119-34-6
EINECS: 204-316-1

Bestellen

Spezifikation Testergebnisse

Appearance Dark brown Crystalline powder
Infrared spectrum Conforms
Melting point 124 to 128 °C
GC ≥98.5 %

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR