DL-α-Phenylalanin 99%

Lieferant: Thermo Fisher Scientific

Synonyme: (±)-2-Amino-3-phenylpropansäure, H-DL-Phe-OH, 3-Phenyl-DL-alanin, DL-Phenylalanin, (±)-2-Amino-3-phenylpropionic acid, (±)-2-Amino-3-phenylpropionsäure

164570250 164571000 164575000
ACRO164570250EA 29.1 EUR
ACRO164570250 ACRO164571000 ACRO164575000
DL-α-Phenylalanin 99%
DL-α-Phenylalanin
Formel: C₉H₁₁NO₂
Molecular Weight: 165.19 g/mol
Siedepunkt: 313
Schmelzpunkt: 266 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064225
CAS-Nummer: 150-30-1
EINECS: 205-756-7
Merck Index: 13,07355

Bestellen

Spezifikation Testergebnisse

Appearance White Powder or crystals
Infrared spectrum Conforms
Loss on drying ≤0.3 %
Sulfated ash ≤0.3 %
Titration with HClO4 98.5 to 101.5 % (on dry substance)
Arsenic (As) ≤3 ppm
Lead (Pb) ≤10 ppm
Total nitrogen ≥9.0 % (on dry substance)
Amino nitrogen ≥3.0 % (on dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR