4-chloro-2-phenylquinoline 99%

Lieferant: Thermo Scientific
Achtung

164870025
ACRO164870025EA 105 EUR
ACRO164870025
4-chloro-2-phenylquinoline 99%
4-chloro-2-phenylquinoline

Appearance: Light yellow Powder

Formel: C₁₅H₁₀ClN
Schmelzpunkt: 62…64 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00160641
CAS-Nummer: 4979-79-7

Bestellen

Spezifikation Testergebnisse

Appearance Light yellow Powder
Infrared spectrum Conforms
Melting point 61 to 64 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR