1-(2-Pyridyl)piperazin 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-(1-Piperazinyl)pyridin
165900250
AC16590.10
AC16590.50
165900050
165901000
ACRO165900250EA
91.7
EUR
ACRO165900250
MAYBAC16590.10
MAYBAC16590.50
ACRO165900050
ACRO165901000
1-(2-Pyridyl)piperazin 98%
1-(2-Pyridyl)piperazin
Formel:
C₉H₁₃N₃ Siedepunkt: 120…122 °C (2 mmHg) Schmelzpunkt: 85 - 87 °C Dichte: 1,07 g/cm³ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006216 CAS-Nummer: 34803-66-2 EINECS: 252-220-3 |
Spezifikation Testergebnisse
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5930 to 1.5960 (20°C, 589 nm) |
GC | ≥97.5 % |
Sulfate (SO4) | ≤0.01 % |
Arsenic (As) | ≤3 ppm |
Lead (Pb) | ≤0.002 % |
Insoluble matter | ≤0.005 % |
Alkalies + earths | ≤0.2 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...