2-(3-Chloropropyl)-2-methyl-1,3-dioxolane 97%

Lieferant: Thermo Fisher Scientific

166040050 166040250
ACRO166040050EA 50.8 EUR
ACRO166040050 ACRO166040250
2-(3-Chloropropyl)-2-methyl-1,3-dioxolane 97%
2-(3-Chloropropyl)-2-methyl-1,3-dioxolane

Appearance: Clear colorless to light yellow Liquid

Formel: C₇H₁₃ClO₂
Siedepunkt: 73…76 °C (7 mmHg)
Dichte: 1,09 g/cm³
Flammpunkt: 72 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003217
CAS-Nummer: 5978-08-5

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4490 to 1.4510 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR