5-Brom-2-thiophencarbaldehyd 97%

Lieferant: Thermo Fisher Scientific
Achtung

166650250 166650050 166651000
ACRO166650250EA 134 EUR
ACRO166650250 ACRO166650050 ACRO166651000
5-Brom-2-thiophencarbaldehyd 97%
5-Brom-2-thiophencarbaldehyd
Formel: C₅H₃BrOS
Dichte: 1,61 g/cm³
Flammpunkt: 98 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00005432
CAS-Nummer: 4701-17-1

Bestellen

Spezifikation Testergebnisse

Appearance Clear yellow to brown Liquid after melting
Infrared spectrum Conforms
Refractive index 1.6380 to 1.6410 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR