1-Benzylpiperidin-3-one hydrochloride hydrate, Hyrdrat 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1-Benzyl-3-piperidone hydrochloride hydrate
166920010
ACRO166920010EA
18.8
EUR
ACRO166920010
1-Benzylpiperidin-3-one hydrochloride hydrate, Hyrdrat 95%
1-Benzylpiperidin-3-one hydrochloride hydrate
Appearance: Cream to light brown Powder
Formel:
C₁₂H₁₅NO.HCl.xH₂O Molecular Weight: 225,72 g/mol (anhy) Schmelzpunkt: 170…176 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00150096 CAS-Nummer: 50606-58-1 UN: 2811 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | Cream to light brown Powder |
Infrared spectrum | Conforms |
Titration Argentometric | 98 to 102 % (on dry substance) |
Water | ≤8 % (K.F.) |
GC | ≥94 % (on free base) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...