2-Adamantanol 98%
Lieferant: Thermo Scientific
Synonyme:
2-Hydroxytricyclo[3.3.1.1(3,7)]decan, Tricyclo[3.3.1.1(3,7)]decan-2-ol, 2-Tricyclo[3.3.1.1(3,7)]decanol
167310050
ACRO167310050EA
66.6
EUR
ACRO167310050
2-Adamantanol 98%
2-Adamantanol
Formel:
C₁₀H₁₆O Molecular Weight: 152,24 g/mol Schmelzpunkt: 258…265 °C Dichte: 1,07 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00074744 CAS-Nummer: 700-57-2 EINECS: 211-846-7 |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 258 to 265 °C (Subl) |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...