Tetrahydro-4H-thiopyran-4-on 99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Tetrahydrothiopyran-4-one
167980050
ACRO167980050EA
87.8
EUR
ACRO167980050
Tetrahydro-4H-thiopyran-4-on 99%
Tetrahydro-4H-thiopyran-4-on
Formel:
C₅H₈OS Molecular Weight: 116,18 g/mol Schmelzpunkt: 62…65 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006660 CAS-Nummer: 1072-72-6 EINECS: 214-015-7 |
Spezifikation Testergebnisse
Appearance | White to light yellow Crystalline solid |
Infrared spectrum | Conforms |
Melting point | 59 to 63 °C |
GC | ≥98.5 % |
Calcium (Ca) | ≤20 ppm |
Copper (Cu) | ≤10 ppm |
Iron (Fe) | ≤20 ppm |
Lead (Pb) | ≤10 ppm |
Magnesium (Mg) | ≤10 ppm |
Nitrogen (N) | ≤20 ppm |
Phosphorus (P) | ≤50 ppm |
Potassium (K) | ≤20 ppm |
Sodium (Na) | ≤50 ppm |
Zinc (Zn) | ≤20 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...