Betahistine dihydrochloride ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: N-Methyl-2-(pyridin-2-yl)ethanamine dihydrochloride

167990010 167990050
ACRO167990010EA 35 EUR
ACRO167990010 ACRO167990050
Betahistine dihydrochloride ≥98%
Betahistine dihydrochloride

  • Bioactive

Formel: C₈H₁₂N₂·2HCl
Molecular Weight: 209.12 g/mol
Schmelzpunkt: 150…154 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00012813
CAS-Nummer: 5579-84-0

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow to beige Crystalline powder
Infrared spectrum Conforms
Melting point 150 to 154 °C
Loss on drying ≤1 % (1 g, 105 °C)
Solubility (5 g in 50 ml water) Clear solution
Titration with NaOH 97.5 to 102.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR