2,6-Dimethylbenzoesäure 99%
Lieferant: Thermo Scientific
|
Achtung
|
169010010
ACRO169010010EA
35.2
EUR
ACRO169010010
2,6-Dimethylbenzoesäure 99%
2,6-Dimethylbenzoic acid
Formel:
C₉H₁₀O₂ Molecular Weight: 150,18 g/mol Siedepunkt: 155 - 160 °C (17 -rr) Schmelzpunkt: 114…117 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002483 CAS-Nummer: 632-46-2 EINECS: 211-177-0 |
Spezifikation Testergebnisse
Appearance | White to cream Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 114 to 117 °C |
Titration with NaOH | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...