Methyl-(triphenylphosphoranyliden)acetat 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Methyl (triphenylphosphoranylidene)acetate, Methyl 2-(triphenylphosphoranylidene)acetate, Methoxycarbonylmethylene-Triphenylphosphorane

169560250 169561000 169565000
ACRO169560250EA 105 EUR
ACRO169560250 ACRO169561000 ACRO169565000
Methyl-(triphenylphosphoranyliden)acetat 98%
Methyl-2-(triphenylphosphoranylidene)acetate

CAS No.: 2605-67-6

Formel: C₂₁H₁₉O₂P
Molecular Weight: 334.36 g/mol
Schmelzpunkt: 164…169 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00008455
CAS-Nummer: 2605-67-6
UN: 2811
ADR: 6.1 III

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 164 to 169 °C
HPLC ≤2 % triphenylphosphine + triphenylphosphine oxide


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR